Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, tissues
Description
The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
- Easy to follow instructions
- Preserves the integrity of the RNA/DNA without degradation
- Choose the method that matches your tissue sample
What you get
The QuantiGene Sample Processing Kit for tissues contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing homogenized tissue from a variety of tissue sources.
Specifications
Specifications
| Content And Storage | • 20 mL Homogenizing Solution, store at 15°C to 30°C • 90 μL Proteinase K (50 μg/μL), store at -20°C |
| Format | Kit |
| Sample Type | Tissue |
| Product Line | QuantiGene |
| Product Type | Sample Processing Kit |
| Quantity | 25 Samples |
| Shipping Condition | Blue Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.