missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control 1137 for Human Serum TIM-1

Product Code. 30001889
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
30001889 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001889 Supplier R&D Systems™ Supplier No. QC229
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunoassay control set for Human T Cell Immunoglobulin Mucin-1 protein

TIM-1 (T cell immunoglobulin and mucin domain 1), also known as KIM-1 and HAVcr1, is expressed on many immune cell types, epithelial cells. It binds to phosphatidylserine (PS), LMIR5/CD300b, TIM-1 (homophilic), TIM-4, IgA, glycoproteins of number of enveloped viruses. Its interaction with PS enables TIM-1 to mediate phagocytosis, clearance of apoptotic cells. TIM-1 signaling costimulates T cell activation, enhances Th2 cytokine production. TIM-1 serves as cellular entry receptor for hepatitis A virus, Ebolavirus and Marburgvirus. Polymorphisms are associated with susceptibility to atopy, autoimmunity, severe hepatitis A virus infection in humans. A soluble form of TIM-1 is elevated in urine during nephropathy.

  • Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes TIM-1
Type Control Set for ELISA Kit
Gene ID (Entrez) 26762 (Human); 171283 (Mouse); 286934 (Rat); 102141332 (Cynomolgus Monkey)
Intended Use For quantitative measurement of T cell immunoglobulin protein levels
Product Line R&D Systems
Product Name Immunoassay Control Set 920
Source Human
Target Gene T Cell Immunoglobulin Mucin-1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.