missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Group 8

Product Code. 30564790
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30564790 1 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30564790 Supplier R&D Systems™ Supplier No. QC23
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / 1 sets
    Save 11.55 €
    5% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Control for Human CXCL6/GCP-2, CXCL8/IL-8, CXCL10/IP-10, CXCL11/I-TAC, Cystatin C, Endoglin/CD105, gp130, IGFBP-3, IL-23, Kallikrein 3/PSA, Pappalysin-1/PAPP-A, RBP4, SCF R/c-kit, and Survivin.

  • For use as quantitative controls for the determination of cytokine concentrations in biological fluids
  • Concentrations have been assigned using R&D Systems’ Quantikine™ kits
  • Controls are prepared in diluted porcine serum with preservatives
  • They contain recombinant human cytokines at low, medium and high concentrations
  • Controls are supplied lyophilized
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Quantikine Immunoassay Control Group 8
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Stability Stable for at least 6 months from date of receipt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.