missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Quantikine Immunoassay Control Set 1203 Human Neuropilin-1

Product Code. 18471910
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18471910 1 Each 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18471910 Supplier R&DSystems™ Supplier No. QC187B
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / 1 sets
    Save 11.55 €
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For use with catalog number DNRP10

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Gene ID (Entrez) 8829
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.