Learn More
R&D Systems™ Quantikine Immunoassay Control Set 582 Human CXCL1/GRO alpha
Description
CXCL1/GRO alpha, also known as KC in mouse and CINC-1 in rat, is member of CXC family of chemokines. Human CXCL1/GRO alpha is 107 amino acids (aa) in length with predicted molecular weight of 11 kDa. The mouse and rat orthologs share 57% , 59% amino acid sequence identity with human protein, respectively. Genome-wide analysis suggests that protein encoded by mouse Cxcl3 gene is homologous to human CXCL1/GRO alpha protein. Human CXCL1/GRO alpha can be cleaved into three isoforms CXCL1/GRO alpha (aa 4-73), CXCL1/GRO alpha (aa 5-73), and CXCL1/GRO alpha (aa 6-73) which show higher activity than full length protein. CXCL1/GRO alpha is secreted by mast cells and macrophages whereupon it acts as chemoattractant for neutrophils.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 2919 (Human); 14825 (Mouse); 81503 (Rat) |
| Intended Use | For quantitative measurement of Interleukin- 2 protein levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 933 |
| Source | Human |
| Target Gene | Interleukin -17 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.