missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 801 Human Insulin C-Pep

Product Code. 30001894
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001894 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001894 Supplier R&D Systems™ Supplier No. QC108
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DICP00

Quantikine Immunoassay Control Set 801 is designed for use as quantitative controls for the determination of human Insulin C-Peptide concentrations in biological fluids.

  • Applications: Control, ELISA
  • Format: Lyophilized
  • Contents: Buffered protein solution with preservatives containing recombinant human proteins at low, medium and high concentrations
  • Research Area: Diabetes
  • Storage: Unreconstituted controls should be stored at 2°C to 8°C and are stable for at least 6 Months from date of receipt
  • For Research Use Only.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Insulin
Sample Type Biological Fluids
Type Immunoassay Control Set
Gene ID (Entrez) 3630
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.