missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 869 for Glucagon

Product Code. 18740938
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18740938 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18740938 Supplier R&D Systems™ Supplier No. QC100
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DGCG0

Quantikine Immunoassay Control Set 869 for Glucagon is a research-grade control for use in ELISA applications. The control is provided as a lyophilized white powder that requires reconstitution with deionized or distilled water.

  • Applications: Control, ELISA
  • Species Reactivity: Multi-Species
  • Related Gene: GCG
  • Storage: Store unopened product at 2°C to 8°C; stable for at least 6 months from date of receipt.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Glucagon
Type Immunoassay Control
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Multi-species
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.