Learn More
Quantikine™ Immunoassay Control Set 886 Human HB-EGF, R&D Systems™
Control Set
Brand:  R&D Systems QC215
Product Code. 18762789
  • 210.00 € / Each
Description
HB-EGF specifically detects HB-EGF in Human samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
Reconstitute each vial with the volume of deionized or distilled water indicated on the product datasheet.
Store the unopened product at 2 - 8° C. Do not use past expiration date.