missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 896 HIV-1 Gag p24

Product Code. 30001918
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001918 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001918 Supplier R&D Systems™ Supplier No. QC221
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

p24 is a major viral core structural protein and its measurement is commonly used as indicator of HIV-1 infection and viral load

For use with catalog number DHP240B

Long Name: Human Immunodeficiency Virus Type 1 GAG/Group-specific Antigen core protein of 24 kDa
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes HIV-1 Gag p24
Type Quantikine Immunoassay Control Set 896 Human HIV-1 Gag p24
Gene ID (Entrez) 155030 (Viral)
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.