missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 913 for Mouse HS TNF-a

Product Code. 30001877
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
30001877 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001877 Supplier R&D Systems™ Supplier No. QC228
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / Pack of 1
    Save 11.55 €
    5% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Immunoassay control set for Mouse Tumor Necrosis Factor protein

Tumor necrosis factor alpha (TNF-α), also known as cachectin and TNFSF2, is prototypic ligand of TNF superfamily. It is pleiotropic molecule that plays central role in inflammation, apoptosis, and immune system development. TNF-α is produced by wide variety of immune and epithelial cell types. Human TNF-α consists of 35 amino acid (aa) cytoplasmic domain, a 21 aa transmembrane segment, and 177 aa extracellular domain (ECD). Within ECD, human TNF-α shares 97% aa sequence identity with rhesus and 71% to 92% with bovine, canine, cotton rat, equine, feline, mouse, porcine, and rat TNF-α. The 26 kDa type 2 transmembrane protein is assembled intracellularly to form noncovalently linked homotrimer. Ligation of this complex induces reverse signaling that promotes lymphocyte costimulation but diminishes monocyte responsiveness. Cleavage of membrane bound TNF-α by TACE/ADAM17 releases 55 kDa soluble trimeric form of TNF-α. TNF-α trimers bind ubiquitous TNF RI and hematopoietic cell-restricted TNF RII, both of which are also expressed as homotrimers. TNF-α regulates lymphoid tissue development through control of apoptosis. It also promotes inflammatory responses by inducing activation of vascular endothelial cells, macrophages. TNF-α is key cytokine in development of several inflammatory disorders. It contributes to development of type 2 diabetes through its effects on insulin resistance and fatty acid metabolism.

  • Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Control Set for ELISA Kit
Gene ID (Entrez) 7124 (Human); 21926 (Mouse); 24835 (Rat); 397086 (Porcine); 280943 (Bovine); 403922 (Canine); 100033834 (Equine); 493755 (Feline); 100009088 (Rabbit)
Intended Use For quantitative measurement of Tumor Necrosis Factor protein levels
Product Name Immunoassay Control Set 913
Source Mouse
Target Gene Tumor Necrosis Factor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.