missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 915 Human Myeloperoxidase

Product Code. 30001932
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001932 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001932 Supplier R&D Systems™ Supplier No. QC211
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / Pack of 1
    Save 11.55 €
    5% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Elevated plasma MPO levels have been associated with a variety of clinical conditions including systemic inflammation, eclampsia, risk of cardiovascular events, vascular endothelial dysfunction, severity of multiple sclerosis, and prospective mortality and oxidative stress during hemodialysis

  • For use as quantitative controls for the determination of cytokine concentrations in biological fluids
  • Concentrations have been assigned using R&D Systems’ Quantikine™ kits
  • Controls are prepared in diluted serum with preservatives
  • They contain recombinant human cytokines at low, medium and high concentrations
  • Controls are supplied lyophilized
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Quantikine Immunoassay Control Set 739 Human Myeloperoxidase
Gene ID (Entrez) 4353 (Human); 17523 (Mouse); 303413 (Rat)
Intended Use Control
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.