missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 1156, Human Total IL-18

Product Code. 30001902
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
30001902 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001902 Supplier R&D Systems™ Supplier No. QC231
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / Pack of 1
    Save 11.55 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Immunoassay control set for Human Interleukin 18 protein levels.

Interleukin 18 (IL-18), also known as interferon-gamma-inducing factor (IGIF) and IL-1γ, is cytokine which shares biologic activities with IL-12 and structural similarities with IL-1 family of proteins. IL-18 was originally cloned from liver cells, has since been shown to be expressed by monocyte/macrophages, osteoblasts and keratinocytes. Caspase-1 (IL-1 beta-converting enzyme) has been implicated in physiological processing of pro-IL-18 to IL-18. Similarly to IL-12, human IL-18 has been shown to enhance NK cell activity in PBMC cultures. Human IL-18 has also been found to induce production IFN-γ and GM-CSF while inhibiting production of IL-10 by PBMC. On enriched human T cells, human IL-18 can enhance Th1 cytokine production and stimulate cell proliferation via IL-2-dependent pathway. In mouse system, IL-18 has been shown to be costimulatory factor for activation of Th1, but not Th2, cells. IL-18 was found to selectively enhance FasL-mediated cytotoxicity of Th1, but not Th0 or Th2, cells. IL-18 has also been shown to induce activated B cells to produce IFN-γ that inhibits IgE production.

  • Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Control Set for ELISA Kit
Gene ID (Entrez) 3606 (Human); 16173 (Mouse); 29197 (Rat); 397057 (Porcine); 574151 (Primate)
Intended Use For quantitative measurement of Interleukin 18 protein levels
Product Name Immunoassay Control Set 919
Source Human
Target Gene Interleukin 18

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.