missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 923 Human GDF-8/Myostatin

Product Code. 30001979
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001979 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001979 Supplier R&D Systems™ Supplier No. QC98
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For use with catalog number DGDF80

The Quantikine Immunoassay Control Set 794 is designed for use with human GDF-8/Myostatin immunoassays.

  • Application: For research use only
  • Composition: Contains ProClin 300, a skin sensitizer
  • Storage: Store in a cool, dry place and keep containers upright
TRUSTED_SUSTAINABILITY

Specifikationer

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes GDF-8 (Myostatin)
Sample Type Biological Fluids
Type Immunoassay Control
Gene ID (Entrez) 2660
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.