missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 925 Human Vitamin D BP

Product Code. 30001875
Change view
Click to view available options
Used for measuring human vitamin D binding proteins
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30001875 Used for measuring human vitamin D binding proteins
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001875 Supplier R&D Systems™ Supplier No. QC185B
  • ONLINE EXCLUSIVE PRICE
    210.00 €
    198.45 € / 1 sets
    Save 11.55 €
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Immunoassay control set for Human Vitamin D binding protein.

Vitamin D binding protein (VitD BP), also known as DBP and Gc-globulin, is 58 kDa glycoprotein that circulates at high concentration in serum and serves as carrier protein for vitamin D. The transport of vitamin D by VitD BP is important for function of wide variety of tissues. VitD BP binds both 25(OH) and hormonally active 1,25(OH)2 forms of vitamin D. VitD BP is primarily expressed in hepatocytes and to lesser extent in kidney. It delivers vitamin D into cells by Megalin-mediated endocytosis. A selectively deglycosylated form of VitD BP known as macrophage activating factor (MAF) blocks angiogenic effects of FGF basic, VEGF, and Angiopoietin 2. VitD BP enhances chemotaxis of monocytes and neutrophils to activated complement component C5a or C5a des Arg (a C-terminally processed form of C5a). The chemotactic cofactor property of VitD BP is eliminated by binding to 1,25(OH)2 vitamin D, but it is not altered by binding to 25(OH) vitamin D or actin.

  • Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Control Set for ELISA Kit
Gene ID (Entrez) 2638 (Human); 14473 (Mouse); 24384 (Rat)
Intended Use For quantitative measurement of vitamin D binding protein levels
Product Name Immunoassay Control Set 925
Source Human
Target Gene Vit D binding protein

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.