missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 933 for Human HS IL-2

Product Code. 30001943
Change view
Click to view available options
Content And Storage:
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
30001943 Store the unopened product at 2°C to 8°C. Do not use past expiration date. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001943 Supplier R&D Systems™ Supplier No. QC238
  • ONLINE EXCLUSIVE PRICE
    223.04 €
    198.45 € / Pack of 1
    Save 24.59 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunoassay control set for Human Interleukin 2 protein levels.

IL-17 family is comprised of at least six proinflammatory cytokines that share conserved cysteine-knot structure but diverge at N-terminus. IL-17 family members are glycoproteins secreted as dimers that induce local cytokine production, recruit granulocytes to sites of inflammation. IL-17 is induced by IL-15, IL-23, mainly in activated CD4+ T cells distinct from Th1 or Th2 cells. IL-17F is most homologous to IL-17, but is induced only by IL-23 in activated monocytes. IL-17B binds IL-17B receptor, but not IL-17 receptor; it is most homologous with IL-17D, which is expressed by resting CD4+ T cells, CD19+ B cells. IL-17E is mainly produced by Th2 cells, recruits eosinophils to lung tissue. IL-17C has very restricted expression pattern but has been detected in adult prostate fetal kidney libraries.

  • Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Type Control Set of Analytes
Gene ID (Entrez) 3558 (Human); 16183 (Mouse); 116562 (Rat); 396868 (Porcine); 280822 (Bovine); 403989 (Canine); 100034204 (Equine); 751114 (Feline); 100302458 (Rabbit)
Product Line R&D Systems
Source Human
Target Gene IL-2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.