missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Allows for the quick and easy determination of chloride in solutions. Macherey Nagel™ Quantofix™ Chloride Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable results within one minute
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store between 2-8°C
Specifications
Specifications
| Color Indicator | Brown to Yellow |
| Format | Strip |
| For Use With (Application) | Determination of Chloride |
| Graduations | 0, 500, 1000, 1500, 2000, ≥ 3000mg/L Cl- |
| Quantity | 100 Pack |
| Storage Requirements | Store between 2−8°C |
| Test Time | 1 min. |
| Type | Semi-qualitative Test Strip |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?