missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Allows for the quick and easy determination of formaldehyde in solutions. Macherey Nagel™ Quantofix™ Formaldehyde Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable result within one minute
- Easily monitor closed cooling or heating circuits
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Specifications
| Color Indicator | Beige to Blue/Violet |
| Format | Strip |
| For Use With (Application) | Determination of Formaldehyde |
| Graduations | 0, 10, 20, 40, 60, 100, 200mg/L |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 1 min. |
| Type | Semi-qualitative Test Strip |
Safety and Handling
Product Identifier
- QUANTOFIX Formaldehyde
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contain: sodium hydroxide solution
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?