missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Molybdenum Test Strip Kits

Product Code. 10063171 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Green
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10063171

Brand: Macherey-Nagel™ 91325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows for the quick and easy determination of molybdenum in solutions. Macherey Nagel™ Quantofix™ Molybdenum Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Rapid: provides reliable results within one minute
  • Excellent for checking coolant and boiler feed water
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator White to Green
Format Strip
For Use With (Application) Determination of Molybdenum
Graduations 0, 5, 20, 50, 100, 250mg/L Mo6+
Quantity 100 Pack
Storage Requirements 30°C
Type Semi-qualitative Test Strip
compliance-icons
Product Identifier
  • QUANTOFIX molybdate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : aminosulphonic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.