missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Peroxide 1000 Test Strip Kits

Product Code. 10233952 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
White to Brown
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10233952 White to Brown Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10233952 Supplier Macherey-Nagel™ Supplier No. 91333

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 1000 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 1000 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Rapid: provides reliable result within 45 seconds
  • Ensures optimal disinfection and verifies potency levels
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator White to Brown
Format Strip
For Use With (Application) Determination of Peroxide 1000
Graduations 0, 50, 150, 300, 500, 800, 1000mg/L H2O2
Quantity 100 Pack
Storage Requirements 30°C
Test Time 15 sec.
Type Semi-qualitative Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.