missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Sulfite Test Strip Kits

Product Code. 10271611 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
White to Salmon
Unit Size:
Pack of 100
Product Code. Color Indicator unitSize
10271611 White to Salmon Pack of 100
1 options
This item is not returnable. View return policy

Product Code. 10271611

Brand: Macherey-Nagel™ 91306

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Allows for the quick and easy determination of sulfite in solutions. Macherey Nagel™ Quantofix™ Sulfite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

Allows for the quick and easy determination of sulfite in solutions. Macherey Nagel™ Quantofix™ Sulfite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Rapid: provides reliable results within 20 seconds
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator White to Salmon
Format Strip
For Use With (Application) Determination of Sulphite
Graduations 0, 10, 25, 50, 100, 250, 500, 1000mg/L SO32-
Quantity 100 Pack
Storage Requirements 30°C
Test Time 20 sec.
Type Semi-qualitative Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.