missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit

Product Code. 18735485 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735485 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735485 Supplier Applied Biosystems™ Supplier No. A32087

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit contains two training plates and a synthetic amplification control to serve as a sample for experiments.

  • A variety of starter kits are available to introduce users to the various applications of the QuantStudio™ 12K Flex OpenArray™ sample block
  • Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments
  • The QuantStudio™ 12K Flex OpenArray™ Vaginal Microbiota Starter Kit contains two training plates and a synthetic amplification control to serve as a sample for experiments
  • This starter kit serves as a training tool for researchers new to this application

The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-probe
Format 384-Well Plate
PCR Method qPCR
Product Line TaqMan
Product Type 12K Flex Vaginal Microbiota Starter Kit
Quantity 1 kit
Shipping Condition Dry Ice
Sample Type DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.