missing translation for 'onlineSavingsMsg'
Learn More

QuantStudio 5 qPCR System

Code produit 15731248 Tous les produits Applied Biosystems Produits
Modifier l'affichage
Click to view available options
Capacity:
384-well 5-20 uL
Conditionnement:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacity Conditionnement
15731248 384-well 5-20 uL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 15731248 Fournisseur ABI Applied Biosystems Code fournisseur a33626
 Restricted Item
Estimated Shipment: 04-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems™ QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems™ QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity. Combined with the AB Assurance* 2 year warranty, the QuantStudio 3 and 5 Real-Time PCR Systems will allow you to use your instruments more effectively and will help reduce risks of downtime by providing a proactive planned maintenance service.

The Applied Biosystems™ QuantStudio™ 3 and 5 Real-Time PCR Systems are the latest additions to our family of high-performance QuantStudio instruments. They provide a modern usability experience similar to mobile devices, allowing you to connect to your data easier than ever before. Therefore, you can access, analyze, and share data anytime, anywhere; and support remote monitoring of your runs, quick analysis of sophisticated data sets, secure storage of your results, and safe sharing of results online with colleagues across campus and around the world with web browser-based software supported by the Thermo Fisher Cloud platform.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems™ QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity. Combined with the AB Assurance* 2 year warranty, the QuantStudio 3 and 5 Real-Time PCR Systems will allow you to use your instruments more effectively and will help reduce risks of downtime by providing a proactive planned maintenance service.

Service Plan

2 Years

Garantie

2 Years

TRUSTED_SUSTAINABILITY

Spécification

Thermal Accuracy 0.25°C
Content And Storage QuantStudio 5 qPCR System (384 well, 0.1mL) and 2 Year Warranty
Thermal Uniformity 0.4°C
Dimensions 50 cm (D) x 27 cm (W) x 40 cm (H)
Capacity 384-well 5-20 uL
Voltage 100/240 V
Display Type Touchscreen
Product Type QuantStudio Real Time PCR System (qPCR)
Height (English) 16 in.
Depth (English) 20 in.
Height (Metric) 40 cm
Width (Metric) 27 cm
Warranty 2 Year
Width (English) 10.5 in.
Depth (Metric) 50 cm
Thermal Range 4°C to 100°C
For Use With (Application) qPCR
Length (Metric) 50 cm
Wattage 960 W
Max. Ramp Rate 6.0°C⁄s
Description QuantStudio 5 qPCR System (384-well, 0.1 mL), Laptop, and 2 Year Warranty
Weight 26 kg (57 lb.)
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.