missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 5 qPCR System

Product Code. 15791238 Shop All Applied Biosystems Products
Change view
Click to view available options
Capacity:
96-well; 10 to 100μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15791238 96-well; 10 to 100μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791238 Supplier Applied Biosystems™ Supplier No. a33621
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems™ QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity. Combined with the AB Assurance* 2 year warranty, the QuantStudio 3 and 5 Real-Time PCR Systems will allow you to use your instruments more effectively and will help reduce risks of downtime by providing a proactive planned maintenance service.

The Applied Biosystems™ QuantStudio™ 3 and 5 Real-Time PCR Systems are the latest additions to our family of high-performance QuantStudio instruments. They provide a modern usability experience similar to mobile devices, allowing you to connect to your data easier than ever before. Therefore, you can access, analyze, and share data anytime, anywhere; and support remote monitoring of your runs, quick analysis of sophisticated data sets, secure storage of your results, and safe sharing of results online with colleagues across campus and around the world with web browser-based software supported by the Thermo Fisher Cloud platform.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems™ QuantStudio Real-Time PCR Systems offer exceptional specificity with the speed and ease–of–use of Cloud connectivity. Combined with the AB Assurance* 2 year warranty, the QuantStudio 3 and 5 Real-Time PCR Systems will allow you to use your instruments more effectively and will help reduce risks of downtime by providing a proactive planned maintenance service.

Service Plan

2 Years

Warranty

2 Years

TRUSTED_SUSTAINABILITY

Specifications

Thermal Accuracy 0.25°C
Content And Storage QuantStudio 5 qPCR System (0.2mL), Laptop, and 2 Year Warranty
Thermal Uniformity 0.4°C
Capacity 96-well; 10 to 100μL
Display Type Touchscreen
Product Type QuantStudio Real Time PCR System (qPCR)
Height (English) 16 in.
Depth (English) 20 in.
Height (Metric) 40 cm
Width (Metric) 27 cm
Warranty 2 Year
Width (English) 10.5 in.
Depth (Metric) 50 cm
Thermal Range 0°C to 100°C
For Use With (Application) qPCR
Length (Metric) 50 cm
Wattage 960 W
Max. Ramp Rate 6.5°C/s
Description QuantStudio 5 qPCR System (0.2 mL) and 2 Year Warranty
Weight 26 kg (57 lb.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.