missing translation for 'onlineSavingsMsg'
Learn More

QuantStudio™ QuantStudio™ 5 Real-Time PCR System, 96-well, 0.2 mL, laptop

Product Code. 15512907 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 system
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15512907 1 system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15512907 Supplier QuantStudio™ Supplier No. A28569
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Laptop Computer

Designed for users who need superior performance, maximum dye versatility, and security options in a real-time PCR system that is affordable and easy to use.

The optimized Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio™ 5 system provides access to your data wherever and whenever you want. Using proven OptiFlex™ technology (featuring 6 decoupled channels and white LED) and featuring six independent VeriFlex™ temperature zones, the QuantStudio™ 5 96-well system enables improved data accuracy and sensitivity for a broad range of genomic applications. The QuantStudio™ 5 system also offers the built-in software features of electronic record security and prevention of unauthorized instrument access to assist with 21 CFR Part 11 compliance.

Features of the QuantStudio™ 5 system with 96-well block include:

  • Proven OptiFlex™ technology (6 decoupled channels/21 filter combinations with white LED) for trusted results
  • User account management, locked workflow, and Pause features for greater control of experiment data
  • Built-in software features to support compliance with 21 CFR Part 11 guidelines
  • Secure cloud-based software provides increased accessibility and storage options
  • Simplified instrument software with interactive touchscreen for greater ease of use
  • 96-well 0.2mL format or 96-well 0.1mL format

Thermo Fisher Cloud:
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Upload data from the instrument or upload your own files from anywhere you have web access. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use.

Easy to use:
The QuantStudio™ 5 Real-Time PCR System is compact in form, factory calibrated and is delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience and the simplified instrument software/pre-optimized protocol templates allow users of any experience level to quickly set up and start a run.

Design and analysis software options:

Option 1: QuantStudio™ Design and Analysis cloud software

  • Supports web browser-based system configuration with PC or MAC computer
  • Streamlined software for improved usability and response time
  • Enables secure access of your data when and where you want through the cloud
  • No software to install, no licenses and no versions to update

Option 2: QuantStudio™ Design and Analysis desktop software

  • Supports traditional co-located computer system configuration on Windows™ 7 OS
  • Streamlined software for improved usability and response time
  • No licenses to purchase

Total control:
With built-in software features to support compliance with 21 CFR Part 11 guidelines, user account management, locked workflow and Pause features, the QuantStudio™ 5 system offers users greater control of experiment data

Multiplex performance results that you can trust:
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex™ technology. In the 96-well system this technology features 6 decoupled excitation and emission filter channels, for a total of 21 filter combinations, with white LED.

Sensitive data analysis:
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

High application versatility:
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Block Format Non-interchangeable
Format 96-well Plate
Content And Storage Laptop Computer
For Use With (Equipment) QuantStudio™
Product Line QuantStudio
Display Type Touchscreen
Shipping Condition Room Temperature
Volume (Metric) Thermal Block Sample 0.2 ml
Quantity 1 system

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.