missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 6 Flex package

Product Code. 15781168 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
PC (Laptop)
PC (Tower)
Block Configurations:
384-well
96-well
96-well FAST
Warranty:
1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year)
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Block Configurations Warranty unitSize
15781168 PC (Laptop) 96-well 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15761158 PC (Laptop) 96-well 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) Each
15761168 PC (Laptop) 96-well FAST 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15771188 PC (Laptop) 384-well 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15791188 PC (Laptop) 384-well 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) Each
15771168 PC (Tower) 96-well 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15731158 PC (Tower) 96-well 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) Each
15751168 PC (Tower) 96-well FAST 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15741158 PC (Tower) 96-well FAST 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) Each
15751188 PC (Tower) 384-well 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
15781188 PC (Tower) 384-well 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 15781168 Supplier Applied Biosystems™ Supplier No. A27181
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Facilitates real-time PCR based applications with an upgradable block selection. With a simplified workflow, intuitive software, and touch-screen interface, the QuantStudio™ 6 Flex system offers exceptional reproducibility at a price you can afford.

  • Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
  • The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block
  • The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage PC (Laptop)
Thermal Uniformity 0.25°C
Capacity 96
Display Type Touchscreen
Height (Metric) 54.7 cm
Width (Metric) 33.8 cm
Warranty 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Depth (Metric) 52.5 cm
Thermal Range 4°C to 99.9°C
Description QuantStudio 6 Flex
Max. Heating Rate (Thermal Block) 96-well: 3.9°C/s
Max. Cooling Rate 3.6°C/s (96-well)
Detection Range Excitation 455 to 672 nm, Emission 505 to 723 nm
Volume (Metric) Thermal Block Sample 0.2 ml (96-well)
Pre-Calibrated Dyes FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
Weight 38 kg
Block Configurations 96-well
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.