missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 7 PRO 0.2mL, 96-well package
GREENER_CHOICE

Product Code. 16277631 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
No PC
PC (Laptop)
PC (Tower)
Warranty:
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Warranty unitSize
16277631 No PC 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
16297631 PC (Laptop) 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
16217641 PC (Tower) 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16277631 Supplier Applied Biosystems™ Supplier No. A44289
Reaching Scientific Milestones Together
Use promo code 27938
View all promotions
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Offers facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.

Features of the QuantStudio 7 Pro Real-Time PCR System include:

  • Easy and convenient hands-free operation with facial authentication and voice commands
  • Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
  • Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
  • Peace of mind with Smart Help; contact Technical Support directly from the touchscreen
  • Reduced downtime with Smart Remote Support; show issues to Technical Support for speedy resolution
  • Application versatility with easily changed blocks and four different block types
  • Increased productivity with automation compatibility for 24/7 operation
  • Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
  • Remotely monitor experiments using the Instrument Connect monitoring app
  • Design and Analysis software compatible with PC and Mac operating systems
  • Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • Provides exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
  • Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage No PC
Thermal Uniformity 0.4°C
Capacity 96
Display Type Touchscreen
Height (Metric) 54.7 cm
Width (Metric) 33.8 cm
Warranty 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Depth (Metric) 52.5 cm
Thermal Range 4°C to 99.9°C
Max. Ramp Rate 6.5°C/s
Description QuantStudio 7 PRO
Excitation Bright White LED
Volume (Metric) Thermal Block Sample 0.2 mL (96-well)
Multiplex 5/6 Targets
Detection CMOS Technology
Pre-Calibrated Dyes FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
Weight 38 kg
Block Configurations 96-well
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.