Learn More
Bellingham Stanley™ Quartz Control Plate Sets
Product Code. 16379125
Change view
Click to view available options
Type:
QCP Full Set: 100°Z, 15°Z, -30°Z
QCP Set A: 100°Z, 15°Z
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
16379125 QCP Set A: 100°Z, 15°Z Each
15977945 QCP Full Set: 100°Z, 15°Z, -30°Z Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16379125 Supplier Bellingham Stanley™ Supplier No. 3430
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Bellingham Stanley™ offer convenient Quartz Control Plate (QCP) sets for verifying and calibrating polarimeters. QCPs are made to the highest standard and supplied with Certificates of Calibration.
  • Convenient Quartz Control Plate (QCP) sets for verifying and calibrating polarimeters
  • Sets available with two or three optical rotation values
  • Inert with relatively low temperature co-efficient
  • Made to the highest standard and supplied with Certificate of Calibration showing traceability to PTB
  • UKAS certified
  • Optional thermal block may be used to provide mechanical contact with the instrument temperature sensor allowing the use of quartz temperature compensation
TRUSTED_SUSTAINABILITY
Specifications
Type QCP Set A: 100°Z, 15°Z
Material Quartz
For Use With (Equipment) Polarimeter