Learn More
Skytec™ Quartz Safety Gloves
Product Code. p-200110046
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274566
Brand: Skytec™ SKY440
Only null left
Add to basket
Add to basket
Ideal for precise operations requiring excellent dexterity and for preventing product contamination. A very supple, stretchable and comfortable mechanical barrier that adapts to every movement.
  • Lightweight with a Breathable PU Palm
  • Abrasion and tear resistant with a secure grip
  • Thin coating facilitates movement
  • Protects from oils and grease
  • Elasticated wrist provides a secure
  • Lint-free
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Abrasion 4,Puncture 2, EN 388:2016 4131X, EN 388:2016 + A1:2018 4131X
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type General Purpose Gloves
Size X-Small
Length (Metric) 260 mm