Learn More
Motic™ Quartz Wedge
Click to view available options
For Use With (Equipment):
BA-310 POL Microscopes
Unit Size:
Each
Description
- Commonly employed for qualitative retardation measurements of petrographic specimens (rock and mineral thin sections) or other birefringent materials whose retardation value falls within the wedge limits.
Specifications
Specifications
| Type | Quartz Wedge |
| For Use With (Equipment) | BA-310 POL Microscopes |