missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits

Product Code. 17399842 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Type:
All Food Types
Fruits and Vegetables with Fats and Waxes
General Fruits and Vegetables
Other Foods
Pigmented Fruits and Vegetables
Pigmented Fruits and Vegetables with Fat
Tube Size:
2 mL
15 mL
Unit Size:
Pack of 100
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Type Tube Size Unit Size
17399842 Other Foods 15 mL Pack of 50
17369832 All Food Types 15 mL Pack of 50
17399832 Fruits and Vegetables with Fats and Waxes 15 mL Pack of 50
17329842 General Fruits and Vegetables 15 mL Pack of 50
17329852 Pigmented Fruits and Vegetables 15 mL Pack of 50
17359852 All Food Types 2 mL Pack of 100
17369852 Fruits and Vegetables with Fats and Waxes 2 mL Pack of 100
17389852 General Fruits and Vegetables 2 mL Pack of 100
17319862 Other Foods 2 mL Pack of 100
17329862 Pigmented Fruits and Vegetables 2 mL Pack of 100
17349862 Pigmented Fruits and Vegetables with Fat 2 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 17399842 Supplier Thermo Scientific™ Supplier No. S215OTHAOACKIT
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Clean-up Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.

Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Clean-up Kits

These kits use dispersive solid phase extraction (dSPE) to purify supernatants obtained during extraction. Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, fruits and vegetables containing fats and waxes, and all other food types.

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
TRUSTED_SUSTAINABILITY

Specifications

Description QuEChERS AOAC Clean-up Kit; 150 mg C18, 900 mg MgSO4 prefilled in 15 mL Tubes; 50/Pk.
Type QuEChERS Clean-up Kit
Includes Salts in Prefilled Tubes
Method QuEChERS sample preparation for AOAC 2007.01 Method
Salt Composition 150 mg C18, 900 mg MgSO4
Sample Type Other Foods
Tube Size 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.