missing translation for 'onlineSavingsMsg'
Learn More

Quercetin Dihydrate, TRC

Product Code. 18040640
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040640 100 mg 100mg
30138631 25 g 25g
30137140 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18040640 Supplier Toronto Research Chemicals Supplier No. TRCQ509500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6151-25-3
Formula Weight 338.0638
InChI Formula InChI=1S/C15H10O7.2H2O/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6;;/h1-5,16-19,21H;2*1H2
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;dihydrate
Molecular Formula C15 H10 O7 . 2 H2 O
Molecular Weight (g/mol) 338.27
Percent Purity >95%
SMILES O.O.OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3
Synonym 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one Dihydrate, Rutoside Trihydrate Imp. C (EP) as Dihydrate, Quercetin Dihydrate
Chemical Name or Material Quercetin dihydrate
Grade HPLC
Show More Show Less

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.