missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quetiapine hemifumarate

Product Code. 15822484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15822484 25 g 25g
15812484 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15822484

Brand: Thermo Scientific Chemicals 462400250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111974-72-2
Molecular Formula C29H33N3O10S
Molecular Weight (g/mol) 615.65
MDL Number MFCD03423782
InChI Key JLWSQVHJLLYDPX-LVEZLNDCSA-N
IUPAC Name bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
SMILES OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 172.0°C to 176.0°C
Color Clear Colorless to Light Yellow
Quantity 25 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in dimethylformamide, slightly soluble in methanol,very slightly soluble, in ether, insoluble in ether and acetonitrile
Formula Weight 441.54
Chemical Name or Material Quetiapine hemifumarate
compliance-icons compliance-icons
Product Identifier
  • Quetiapine hemifumarate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H370-Causes damage to organs.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Quetiapine fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.