missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine hemifumarate, MedChemExpress

Product Code. 16442039
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16442039 1 g 1g
16400768 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 16442039

Brand: Medchem Express HYB0031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quetiapine hemifumarate is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine hemifumarate is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine hemifumarate has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Quetiapine hemifumarate is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine hemifumarate is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine hemifumarate has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Specifications

Chemical Name or Material Quetiapine hemifumarate
CAS 111974-72-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27N3O4S
Quantity 1 g
Solubility Information DMSO : 50 mg/mL (113.24 mM; Need ultrasonic) ∣H2O : 1.25 mg/mL (2.83 mM; Need ultrasonic)
SMILES OCCOCCN(CC1)CCN1C2=NC(C=CC=C3)=C3SC4=C2C=CC=C4.O=C(O)/C=C/C(O)=O.[0.5]
Molecular Weight (g/mol) 441.54
Formula Weight 441.54
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.