missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Hydroxy Impurity Dihydrochloride Salt, TRC

Product Code. 30133762
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
50 mg
Unit Size:
2.5mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133762 50 mg 50mg
30135585 25 mg 25mg
30009072 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133762 Supplier Toronto Research Chemicals Supplier No. TRCQ51001050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329218-14-6
Molecular Formula C19 H21 N3 O S . 2 Cl H
Molecular Weight (g/mol) 412.38
Synonym 1-Piperazineethanol, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, hydrochloride (1:2), 1-Piperazineethanol, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, dihydrochloride (9CI), 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol dihydrochloride, Quetiapine Hydroxy Impurity Dihydrochloride Salt
IUPAC Name 2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethanol;dihydrochloride
SMILES Cl.Cl.OCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H21N3OS.2ClH/c23-14-13-21-9-11-22(12-10-21)19-15-5-1-3-7-17(15)24-18-8-4-2-6-16(18)20-19;;/h1-8,23H,9-14H2;2*1H
Formula Weight 411.0939
Chemical Name or Material 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.