missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Quick Wipes

Product Code. 15318964
Click to view available options
Packaging Quantity:
12 x 120 Wipes
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 15318964

Brand: Laboratoires Anios™ 2333421BZ

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Laboratoires Anios™ Quick Wipes are a convenient, fast-acting disinfectant wipe for use with non-immersible medical devices.

  • Disinfectant wipes for non-immersible medical devices
  • Ethanol-based
  • Wide spectrum in short time
  • Aldehyde- and colorant-free formula
  • Pleasant fragrance
  • Suitable for use with previously cleaned, alcohol-resistant, non-immersible medical devices (stethoscopes, cables and connectors, pressure sensors, blood sugar testers, etc.)
  • No need to rinse unless the medical device comes into contact with the skin or mucosa
  • Active against bacteria, mycobacteria, yeasts, molds, viruses

Specifications

Product Type Disinfectant
Construction Cellulose, viscose, acrylic binder
Disposable Single-use
Dimensions (L x W) 130 x 190 mm
For Use With (Application) Non-immersible, previously cleaned, alcohol-resistant medical devices (stethoscopes, cables and connectors, pressure sensors, blood sugar testers, etc.)
Packaging Quantity 12 x 120 Wipes
Saturation Solution Ethanol (55%), quaternary ammonium propionate, fragrance
Certifications/Compliance (Europe) Bacteria: EN 1040, EN 13727, EN 14561, EN 13697. MRSA according to EN 14561Mycobacteria: Mycobacterium terrae : EN 14348, EN 14563Mycobacterium avium : EN 14348, EN 1456Yeasts: Candida albicans : EN 1275, EN 13624, EN 14562,EN 13697Molds: Tricophyton mentagrophytes according to EN 14562Aspergillus niger : EN 1275, EN 13624, EN 14562,EN 13697Viruses: Adenovirus, HIV-1, PRV (surrogate of HBV), BVDV (surrogate of HCV), Vaccinia virus, Herpes-virus, Coronavirus, Rotavirus, Influenza virus A (H1N1), Calicivirus FelinMNV according to EN 14476
compliance-icons compliance-icons
Product Identifier
  • ANIOS QUICK WIPES
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501-Dispose of contents/container to as hazardous waste..
Supplemental information
  • MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL, N,N-DIDECYL-N-METHYL-POLY(OXYETHYL)AMMONIUM PROPIONATE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.