Learn More
Quickfit™ Allihn Condenser with Ground Cone
Click to view available options
Standard Taper Joint No:
19/26
24/29
29/32
34/35
40/38
50/42
Unit Size:
Each
Description
Ideal for reflux applications.
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Specifications
| For Use With (Application) | Ideal for reflux work |
| Length (Metric) Jacket | 365 mm |
| Length (Metric) | 365 mm |
| Standard Taper Joint No | 40/38 |
| Material | Borosilicate Glass |
| Type | Allihn Condenser |