Learn More
Quickfit™ Borosilicate Glass Dufton Fractionating Column
Product Code. 15517820
Click to view available options
Length (Metric):
270mm
290mm
300mm
425mm
Unit Size:
Each
Pack of 5
Product Code. 15517820
Brand: Quickfit™ FC2/23
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Efficient fractionating column designed with an an accurately fitted Dufton glass spiral. Quickfit™ Borosilicate Glass Dufton Fractionating Column can be disassembled for cleaning.
  • Glass column with accurately fitted Dufton glass spiral
  • Good efficiency - liquid spiralling down column meets ascending vapour
  • Spiral can be removed for cleaning
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 300mm
Type Dufton Fractionating Column
Material Borosilicate Glass
Certifications/Compliance ISO 3585