missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Florentine Flask
Allows easy removal of solid residues
Brand: Quickfit FD50/4RE
70.40 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 50
Additional Details : Weight : 0.14400kg
Description
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
50 mL | |
Borosilicate Glass | |
94 mm |