Learn More
Quickfit™ Borosilicate Glass Florentine Flask
Allows easy removal of solid residues
41.45 € - 338.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11351895
|
Quickfit
FD100/3 |
100 mL |
41.45 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
12613182
|
Quickfit
FD50/4RE |
50 mL |
74.10 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15547820
|
Quickfit
FD1L/3 |
1000 mL |
338.00 €
Pack of 5 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15557820
|
Quickfit
FD250/3 |
250 mL |
255.00 €
Pack of 5 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
15567820
|
Quickfit
FD500/3 |
500 mL |
295.00 €
Pack of 5 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
Description
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
Borosilicate Glass |