missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Florentine Flask

Product Code. 15547820
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 15547820

Brand: Quickfit FD1L/3

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows easy removal of solid residues

  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 204 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.