missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Borosilicate Glass Florentine Flask
Allows easy removal of solid residues
Brand: Quickfit FD250/3
Code : NEW
Additional Details : Weight : 0.14000kg
Description
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
250 mL | |
Borosilicate Glass | |
148 mm |