Learn More
Quickfit™ Borosilicate Glass Still Head Adapter
Product Code. 12098483
Click to view available options
Standard Taper Joint:
Cone: 14/23 (Condenser), 14/23 (Flask); Socket: 14/23
Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 14/23
Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 19/26
Cone: 19/26 (Condenser), 24/29 (Flask); Socket: 14/23
Cone: 24/29 (Condenser), 24/29 (Flask); Socket: 14/23
Cone: 29/32 (Condenser), 29/32 (Flask); Socket: 14/23
Cone: 34/35 (Condenser), 34/35 (Flask); Socket: 14/23
Unit Size:
Each
Pack of 5
Product Code. 12098483
Brand: Quickfit SH4/44
Edge
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
  • Used in distillation assemblies for connecting flasks to condenser
  • Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Connecting flask and condenser
Type Still Head Adapter
Material Borosilicate Glass
Standard Taper Joint Cone: 29/32 (Condenser), 29/32 (Flask); Socket: 14/23