Learn More
Quickfit™ Polyacetal Clamp for Spherical Joints
Click to view available options
Color:
Dark Red
Light Orange
Violet
Unit Size:
Pack of 5
Description
- Manufactured from polyacetal
- Can considerably reduce the need for clamping
- Easy to fit and remove
- Can be used at temperatures up to 80°C
- Clips are color coded for easy identification
Specifications
Specifications
| Quantity | 5/Pk. |
| Type | Joint Clamp |
| Material | Polyacetal |
| Color | Violet |