Learn More
Quickfit™ Polyacetal Clamp for Spherical Joints
Product Code. p-8002683
Click to view available options
Color:
Dark Red
Light Orange
Violet
Unit Size:
Pack of 5
Product Code. 12613032
Brand: Quickfit KS13
Only null left
Add to basket
Add to basket
For retaining glassware having Quickfit™ spherical joints
  • Manufactured from polyacetal
  • Can considerably reduce the need for clamping
  • Easy to fit and remove
  • Can be used at temperatures up to 80°C
  • Clips are color coded for easy identification
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5/Pk.
Type Joint Clamp
Material Polyacetal
Color Violet