Learn More
Quickfit™ Right Angle Tubing Adapter
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
Description
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
- With right angle connection for attachment of rubber hose etc
- Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
Specifications
| Cone Size | 29/32 |
| For Use With (Equipment) | Flexible tubing, Rubber hose |
| Includes | Detachable Plastic screw thread connector |
| Material | Borosilicate Glass |
| Product Type | Right angle tubing adapter |