Learn More
Quickfit™ Right Angle Tubing Adapter
Product Code. 12098361
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
Product Code. 12098361
Brand: Quickfit MF10/1B/SC
Only null left
Add to basket
Edge
Add to basket
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
  • With right angle connection for attachment of rubber hose etc
  • Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 14/23
For Use With (Equipment) Flexible tubing, Rubber hose
Includes Detachable Plastic screw thread connector
Material Borosilicate Glass
Product Type Right angle tubing adapter