Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
64.60 € - 101.00 €
Specifications
For Use With (Application) | Ideal for securing thermometers, gas inlet tubes |
---|---|
Includes | Plastic cap, Silicone Rubber ring and PTFE washer |
Material | Borosilicate Glass |
Product Type | Cone screw thread adapter |
Product Code | Brand | Thread Size | Cone Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Cone Size | Price | Quantity & Availability | ||||
10821900
|
Quickfit
ST52/13 |
13 | 19/26 |
66.70 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10840580
|
Quickfit
ST54/24 |
24 | 29/32 |
95.45 €
Pack of 2 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10654969
|
Quickfit
ST52/18 |
18 | 19/26 |
64.60 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10187988
|
Quickfit
ST51/13 |
13 | 14/23 |
83.75 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10684009
|
Quickfit
ST53/18 |
18 | 24/29 |
101.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10290219
|
Quickfit
ST51/18 |
18 | 14/23 |
75.70 €
Pack of 2 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
10168138
|
Quickfit
ST53/13 |
13 | 24/29 |
75.55 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10484520
|
Quickfit
ST53/24 |
24 | 24/29 |
84.65 €
Pack of 2 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10426060
|
Quickfit
ST52/24 |
24 | 19/26 |
74.05 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
10494520
|
Quickfit
ST54/13 |
13 | 29/32 |
84.65 €
Pack of 2 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
10870390
|
Quickfit
ST54/18 |
18 | 29/32 |
84.65 €
Pack of 2 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
- With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
- Ideal for securing thermometers, gas inlet tubes, etc
Specifications
Ideal for securing thermometers, gas inlet tubes | |
Plastic cap, Silicone Rubber ring and PTFE washer | |
Borosilicate Glass | |
Cone screw thread adapter |