Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Click to view available options
Standard Taper Joint:
7/16
Standard; 10/19
Standard; 12/21
Standard; 14/23
Standard; 19/26
Standard; 24/29
Standard; 29/32
Standard; 34/35
Standard; 45/40
Unit Size:
Pack of 10
Pack of 2
Pack of 5
Description
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
- Conforms to the standard tapers as outlined in ISO 383
- Borosilicate glass for chemical durability and high temperature performance
- Hollow-blown construction to reduce weight
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Specifications
Specifications
| For Use With (Application) | Chemical durability and high temperature performance |
| For Use With (Equipment) | Quickfit™ jointed ware and other Glassware having standard ground Glass sockets |
| Standard Taper Joint | Standard; 24/29 |
| Material | Borosilicate Glass |
| Product Type | Hollow Stopper |