Learn More
Quickfit™ Swan Neck Adapter
Product Code. 12435306
Change view
Click to view available options
Cone Size:
19/26
24/29
Unit Size:
Each
Product Code. Cone Size unitSize
12435306 19/26 Each
12071603 24/29 Each
2 options
Product Code. 12435306
Brand: Quickfit MA6/22
Only null left
Add to basket
Add to basket
Has screwthread joint to accept thermometers and tubes
  • Available in different socket, cone, and thread sizes
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 19/26
For Use With (Application) Ideal for securing thermometers, gas inlet tubes
Material Borosilicate Glass
Product Type Swan neck adapter
Thread Size 13