Learn More
Tocris Bioscience™ Quin C1
GREENER_CHOICE
Potent and selective FPR2 agonist
Brand:  Tocris Bioscience™ 5583/50
Encompass
Product Code. 15756946
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Quin C1 is a potent and selective FPR2 agonist (EC50 = 15 nM). Induces Ca2+ mobilization in FPR2 but not FPR1-transfected mast cells. Induces neutrophil chemotaxis and degranulation in vitro. Reduces neutrophil and lymphocyte counts in bronchoalveolar lavage fluid in a mouse lung injury model. Anti-inflammatory.
Specifications
Specifications
4-Butoxy-N-[1,4-dihydro-2-(4-methoxyphenyl)-4-oxo-3(2H)-quinazolinyl]benzamide
786706-21-6